EMAIL THIS PAGE TO A FRIEND
Clinical chemistry
Extraction of Kober chromogen into tetrachloroethane containing trichloroacetic acid in determination of urinary estrogens during pregnancy.
PMID 719884
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W241407
Ethyl acetate, ≥99%, FCC, FG
C4H8O2