EMAIL THIS PAGE TO A FRIEND
Journal of theoretical biology
Biophysical studies on molecular mechanism of abortificient action of prostaglandins-IV. Conformation energy calculation of PGA1, PGB1 and PGE1.
PMID 7200559