EMAIL THIS PAGE TO A FRIEND
Experimental cell research
In vitro comparison of embryonic myoblasts and myogenic cells isolated from regenerating adult rat skeletal muscle.
PMID 7200905
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A25120 DEAE–Sephadex® A-25 chloride form