EMAIL THIS PAGE TO A FRIEND
Journal of bacteriology
Soluble aldehyde dehydrogenase and metabolism of aldehydes by soybean bacteroids.
PMID 7202001
Abstract
A soluble aldehyde dehydrogenase (EC 1.2.1.3) was partially purified from Rhizobium japonicum bacteroids and from free-living R. japonicum 61A76. The enzyme was activated by NAD+, NADH, and dithiothreitol, and it reduced NAD(P)+. Acetaldehyde, propionaldehyde, butyraldehyde, benzaldehyde, and succinic semialdehyde were substrates. The Km for straight-chain aldehydes decreased with increasing carbon chain length. The aldehyde dehydrogenase was inhibited by 6-cyanopurine, but not by metronidazole. These compounds inhibited acetylene reduction, but not respiration, by isolated bacteroids.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
852503
6-Cyanopurine, 97%
C6H3N5
C5392
6-Cyanopurine, ~90%
C6H3N5