EMAIL THIS PAGE TO A FRIEND
Rinsho byori. The Japanese journal of clinical pathology
[A colorimetric method using 2,6-dichloroquinonechloroimide as color reagent for the determination of alkaline phosphatase in serum (author's transl)].
PMID 7206261