EMAIL THIS PAGE TO A FRIEND
Therapie
[Effects of liver disease and age on metabolism of trimethoxybenzene (author's transl)].
PMID 7209856
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
138827
1,3,5-Trimethoxybenzene, ReagentPlus®, ≥99%
C9H12O3