EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Simultaneous demonstration of phagocytosis-connected oxygen consumption and corresponding NAD(P)H oxidase activity: direct evidence for NADPH as the predominant electron donor to oxygen in phagocytizing human neutrophils.
PMID 7225119