EMAIL THIS PAGE TO A FRIEND
International journal of peptide and protein research
Preparation and conformational study of clupeine fragments.
PMID 7228496
Abstract
Fragments of clupeines, YI, YII, and Z of divergent chain length and different amino acid composition were prepared by digestion with thermolysin and a mixture of carboxypeptidases A and B, and their conformational preferences examined as a function of pH, added salts, presence of a helix-supporting solvent, and temperature. All these highly basic oligopeptides adopt an essentially unordered conformation in aqueous solution. 2-Chloroethanol supports in various amounts the onset of the right-handed alpha-helical form in the carboxy-peptidase fragments.
Related Materials