EMAIL THIS PAGE TO A FRIEND
International journal of nuclear medicine and biology
Assessment of hepatic tissue blood flow with 133Xe clearance technique.
PMID 7228521
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
293768
1-Methyluracil, 99%
C5H6N2O2