EMAIL THIS PAGE TO A FRIEND
Die Medizinische Welt
[Treatment of extrapyramidal tremors in the aged].
PMID 7231137
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M1808000 Metixene hydrochloride, European Pharmacopoeia (EP) Reference Standard
C20H23NSl · HCl · H2O