EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
A comparison of in vivo and in vitro (tissue explant) techniques: metabolic profile of ethylcyclohexane in rat and dog.
PMID 7236905
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E19154
Ethylcyclohexane, ≥99%
C8H16