EMAIL THIS PAGE TO A FRIEND
Gigiena truda i professional'nye zabolevaniia
[Toxicity, hazard and nature of the biological action of methylcyclopentadiene dimer].
PMID 7239290
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
129828
Methylcyclopentadiene dimer, 93%
C12H16