EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
The mobility of cholesteryl esters in native and reconstituted low density lipoprotein as monitored by nuclear magnetic resonance spectroscopy.
PMID 7240142
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8753
Cholesteryl arachidonate, ≥95% (HPLC; detection at 205 nm), viscous liquid
C47H76O2