EMAIL THIS PAGE TO A FRIEND
Die Naturwissenschaften
Different inhibition pattern of the nucleic acid metabolism after in vitro treatment with titanocene and vanadocene dichlorides.
PMID 7242686
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
334642
Bis(cyclopentadienyl)vanadium(IV) dichloride, 95%
C10H10Cl2V