EMAIL THIS PAGE TO A FRIEND
Revista medico-chirurgicala a Societatii de Medici si Naturalisti din Iasi
[Determination of the carcinogenic risk of diaminodibenzyl (DADB)].
PMID 7244477
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32784
4,4′-Ethylenedianiline, technical, ≥95% (NT)
C14H16N2