EMAIL THIS PAGE TO A FRIEND
Toxicology letters
Elimination pattern and biodistribution of [14C]chlorferron in rats.
PMID 7245230
Abstract
Administration of [14C]chlorferron in a single oral dose of 0.5 and 20 mg/kg body weight to female rats resulted in a urinary excretion of greater than 74% of the given dose during the first 24 h. Approx. 8% of the dose was eliminated in faeces within 7 days. 7 days after dosing, very low levels of [14C]chlorferron-derived residues were detected in all analyzed organs. These findings indicated that chlorferron was absorbed from the gastro-intestinal tract in appreciable quantities, but was rapidly excreted mainly via the urine with small amounts only in the faeces.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
640611
3-Chloro-7-hydroxy-4-methylcoumarin, 97%
C10H7ClO3