EMAIL THIS PAGE TO A FRIEND
FEBS letters
Herbicides and fungicides stimulate Ca2+ efflux from rat liver mitochondria.
PMID 7250372
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
03992
Amiprofos methyl, PESTANAL®, analytical standard
C11H17N2O4PS