EMAIL THIS PAGE TO A FRIEND
Gigiena truda i professional'nye zabolevaniia
[Experimental studies on calcium borate toxicity].
PMID 7250732
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11618
Calcium metaborate, technical, 39-44% B2O3 basis, 31-37% CaO basis, powder
B2CaO4 · 2H2O