EMAIL THIS PAGE TO A FRIEND
The Journal of antibiotics
The structure of siomycin-D1, peptide antibiotic isolated from Streptomyces sioyaensis.
PMID 7251492
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S6076
Siomycin A, from Streptomyces sioyaensis, ≥98% (HPLC)
C71H81N19O18S5