EMAIL THIS PAGE TO A FRIEND
Journal of biochemistry
Bindings of monodispersed n-alkylphosphorylcholines to cobra venom phospholipases A2.
PMID 7251577
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
38544
O-(Octylphosphoryl)choline solution, 1 M in H2O
C13H30NO4P