EMAIL THIS PAGE TO A FRIEND
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society
Heavy metal intensification of DAB-based HRP reaction product.
PMID 7252134