EMAIL THIS PAGE TO A FRIEND
The Journal of membrane biology
The effect of electrical gradients on current fluctuations and impedance recorded from Necturus gallbladder.
PMID 7253010
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T45802
2,4,6-Triaminopyrimidine, 97%
C4H7N5