EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Measurement of exposure to xylenes by separate determination of m- and p-methylhippuric acids in urine.
PMID 7263793
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
328022
4-Methylhippuric acid, 98%
C10H11NO3