EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical sciences
Studies on drug metabolism by use of isotopes XXVI: Determination of urinary metabolites of rutin in humans.
PMID 7264927
Abstract
Determination of urinary metabolites of orally administered rutin and rutin-2',5',6'-d3 in humans was carried out by TLC and GLC-mass spectrometry. In human urine, 3-hydroxyphenylacetic acid, 3-methoxy-4-hydroxyphenylacetic acid, 3,4-dihydroxyphenylacetic acid, 3,4-dihydroxytoluene, and beta-m-hydroxyphenylhydracrylic acid were identified as rutin metabolites. Unchanged rutin and quercetin were not present in urine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H49901
3-Hydroxyphenylacetic acid, ≥99%
C8H8O3
M34200
4-Methylcatechol, ≥95%
C7H8O2