EMAIL THIS PAGE TO A FRIEND
Die Pharmazie
[Gas chromatographic determination of chlorpropham, metobromuron and chlorbromuron residues in drugs of the East German Pharmacopoeia, 2d edition].
PMID 7267691