EMAIL THIS PAGE TO A FRIEND
Acta pharmaceutica Hungarica
[Pharmaco-analytical application of tetrabromophenolphthalein ethyl ester indicator dye. III. Interaction between alkaloids and TBPE].
PMID 7270183