EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Effects of acetylated polyamines on ornithine decarboxylase in rat HTC cells.
PMID 7271809
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8784
N-Acetylputrescine hydrochloride, ≥98% (TLC)
C6H14N2O · HCl