EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
Increased inhibition of hepatic uroporphyrinogen decarboxylase by hexachlorobenzene in male rats given the oestrogenic drugs diethylstilboestrol and chlorotrianisene.
PMID 7271874