EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
A comparison of biphenyl 4-hydroxylation and 4-methoxybiphenyl O-demethylation in rat liver microsomes.
PMID 7271882
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
374776
4-Methoxybiphenyl, 97%
C13H12O