EMAIL THIS PAGE TO A FRIEND
Vrachebnoe delo
[Ultrastructural bases for cardiovascular insufficiency in experimental chronic pneumonia].
PMID 7271956
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9140
Convallatoxin, ≥65%
C29H42O10