EMAIL THIS PAGE TO A FRIEND
Indian journal of biochemistry & biophysics
In vivo & in vitro inhibition of mung bean superoxide dismutase by cupferron.
PMID 7275160
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
675636
Cupferron, 97%, reagent grade
C6H9N3O2