EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Bromoacetylcholine as an affinity label of the acetylcholine receptor from Torpedo californica.
PMID 728154
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B121
Bromoacetylcholine bromide
C7H15BrNO2 · Br