EMAIL THIS PAGE TO A FRIEND
Chemotherapy
Energy-linked condensed-orthodox ultrastructural transformations in mitochondria.
PMID 7285635
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56915
Indazole-3-carboxylic acid, ≥97.0% (HPLC)
C8H6N2O2