EMAIL THIS PAGE TO A FRIEND
Farmakologiia i toksikologiia
[Changes in the toxicity of certain cardiovascular drugs as a result of hyperbaric oxygenation].
PMID 7286189
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9140
Convallatoxin, ≥65%
C29H42O10