EMAIL THIS PAGE TO A FRIEND
Folia medica Cracoviensia
[Effect of anticoagulants of the 4-hydroxycoumarin derivative group on aminotransferase activity in vitro and in vivo].
PMID 7286839
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I2002
1,3-Indandione, 97%
C9H6O2