EMAIL THIS PAGE TO A FRIEND
Radiobiologiia
[Postradiation DNA repair in mammalian cells in combined exposure to hyperthermia, 8-bromocaffeine and actinomycin D].
PMID 7291482
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
775789
8-Bromocaffeine, 98%
C8H9BrN4O2