EMAIL THIS PAGE TO A FRIEND
Anesteziologiia i reanimatologiia
[Comparative evaluation of the effect of digoxin, strophanthin, isolanid and corglycon on the indices of central hemodynamics, vascular tonus, circulating blood volume and myocardial contractility in acute heart failure].
PMID 7294441
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9140
Convallatoxin, ≥65%
C29H42O10