EMAIL THIS PAGE TO A FRIEND
Experientia
Antifertility activity of trimethylphosphate in male rats.
PMID 7297669
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
241024
Trimethyl phosphate, ≥99%
C3H9O4P
132195
Trimethyl phosphate, 97%
C3H9O4P