EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical sciences
Synthesis and evaluation of guanazole prodrugs as antineoplastic agents.
PMID 7299678
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D26202
3,5-Diamino-1,2,4-triazole, 98%
C2H5N5