EMAIL THIS PAGE TO A FRIEND
Yao xue xue bao = Acta pharmaceutica Sinica
[Studies on antiarrhythmics. I. Synthesis of 3,4,5,-trimethoxybenzamide derivatives (author's transl)].
PMID 7304186
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
567612
3,4,5-Trimethoxybenzamide, 97%
C10H13NO4