EMAIL THIS PAGE TO A FRIEND
Bratislavske lekarske listy
[Effect of 2-phenyl-4,5-dichlor-3-pyridazinone upon respiration and immunoglobulin levels (author's transl)].
PMID 7306834
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
176842
1-Phenyl-4,5-dichloro-6-pyridazone, ≥99%
C10H6Cl2N2O