EMAIL THIS PAGE TO A FRIEND
Developments in toxicology and environmental science
Hexachlorobenzene (HCB) and some of its sulfur-containing metabolites: tissue retention and excretion routes in mice.
PMID 7308072