EMAIL THIS PAGE TO A FRIEND
European journal of cancer
Tumor inhibition by metallocenes: activity against leukemias and detection of the systemic effect.
PMID 7308264
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
334642
Bis(cyclopentadienyl)vanadium(IV) dichloride, 95%
C10H10Cl2V