EMAIL THIS PAGE TO A FRIEND
National Cancer Institute monograph
Current investigational drugs of interest in the Chemotherapy Program of the National Cancer Institute.
PMID 73134
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D26202
3,5-Diamino-1,2,4-triazole, 98%
C2H5N5