EMAIL THIS PAGE TO A FRIEND
Roczniki Panstwowego Zakladu Higieny
[Determination of dodine in selected plant material].
PMID 7313466
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45466
Dodin, PESTANAL®, analytical standard
C15H33N3O2
N11840
Dodin, technical grade
C15H33N3O2