EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Measurement of ionized magnesium in biological fluids.
PMID 7316161
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
201723
Plasmocorinth B, Dye content 60 %
C16H9ClN2Na2O9S2