EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Affinity chromatographic isolation of calmodulin from bovine-brain acetone powder.
PMID 7316162
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B0508 Brain acetone powder bovine