EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Gas--liquid chromatography and mass spectrometry of methylated and acetylated methyl glycosides. Application to the structural analysis of glycoprotein glycans.
PMID 7316180
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M0257
Methyl N-acetyl-α-D-glucosaminide
C9H17NO6