EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
The antagonism of the toxicity of hypoglycin by glycine.
PMID 7317076
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
63978
(RS)-(Methylenecyclopropyl)acetic acid, analytical standard
C6H8O2