EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Production and assay of antibodies to acetylcholine receptors.
PMID 7321891
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56834 Immunoglobulin G from human serum, ≥95% (GE)